Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diphenylpropionic acid, 97%, Thermo Scientific™

Catalog No. AC117410250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC117410250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC117410250 Supplier Thermo Scientific Chemicals Supplier No. 117410250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5558-66-7
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C15H14O2
Linear Formula CH3C(C6H5)2CO2H
Quantity 25g
MDL Number MFCD00004189
Beilstein 09, II, 474
Synonym 2,2-diphenylpropionic acid, 2,2-diphenylpropionicacid, a,a-diphenylpropionic acid, alpha,alpha-diphenylpropionic acid, acmc-209lnv, cambridge id 7170182, 2,2-diphenyl propanoic acid, 2,2-diphenyl propionic acid, 2,2-di phenyl propanoic acid, #
InChI Key ODELFXJUOVNEFZ-UHFFFAOYSA-N
SMILES CC(C1=CC=CC=C1)(C2=CC=CC=C2)C(=O)O
IUPAC Name 2,2-diphenylpropanoic acid
Molecular Weight (g/mol) 226.275
PubChem CID 79676
Formula Weight 226.27
Percent Purity ≥96.0%
Chemical Name or Material 2, 2-Diphenylpropionic acid, 97%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.