Learn More
Medchemexpress LLC 2,2'-Dipyridyl disulfide | 2127-03-9 | 100.0% | 220.32 | 25 G
SDP
Catalog No. 5000366089
Encompass_Preferred
2,2'-Dipyridyl disulfide is a chemical reagent commonly used in peptide chemistry and for the determination of sulfhydryl groups. It facilitates oxidation-reduction condensations to form peptide bonds and is employed in coupling reactions to create disulfide-linked heterodimers. This compound can significantly accelerate the formation of intramolecular S-S bonds when added to air-oxidized mixtures of cysteine-containing peptides.
  • Useful reagent for sulfhydryl group determination.
  • Employed in oxidation-reduction condensations for peptide bond formation.
  • Used in coupling reactions to form disulfide-linked heterodimers.
  • Accelerates intramolecular S-S bond formation in cysteine-containing peptides.
Catalog No. 50-003-66089 Supplier Medchemexpress LLC Supplier No. 25GHYY1666
2,2'-Dipyridyl disulfide is a chemical reagent commonly used in peptide chemistry and for the determination of sulfhydryl groups. It facilitates oxidation-reduction condensations to form peptide bonds and is employed in coupling reactions to create disulfide-linked heterodimers. This compound can significantly accelerate the formation of intramolecular S-S bonds when added to air-oxidized mixtures of cysteine-containing peptides.
  • Useful reagent for sulfhydryl group determination.
  • Employed in oxidation-reduction condensations for peptide bond formation.
  • Used in coupling reactions to form disulfide-linked heterodimers.
  • Accelerates intramolecular S-S bond formation in cysteine-containing peptides.