Learn More
Medchemexpress LLC 2,2'-Dipyridyl disulfide | 2127-03-9 | 99.9% | 220.31 | 10 MM * 1 ML
SDP
Catalog No. 5000365393
Encompass_Preferred
2,2'-Dipyridyl disulfide is a useful reagent for the determination of sulfhydryl groups. It is a common reagent in peptide chemistry, frequently used in oxidation-reduction condensations to form peptide bonds or in coupling reactions to form disulfide-linked heterodimers. It can dramatically accelerate the formation of intramolecular S-S bonds when added to an air-oxidized mixture of cysteine-containing peptides.
  • Useful reagent for sulfhydryl group determination
  • Common reagent in peptide chemistry
  • Used in oxidation-reduction condensations to form peptide bonds
  • Used in coupling reactions to form disulfide-linked heterodimers
  • Accelerates intramolecular S-S bond formation in cysteine-containing peptides
Catalog No. 50-003-65393 Supplier Medchemexpress LLC Supplier No. 10MM/1MLHYY1666
2,2'-Dipyridyl disulfide is a useful reagent for the determination of sulfhydryl groups. It is a common reagent in peptide chemistry, frequently used in oxidation-reduction condensations to form peptide bonds or in coupling reactions to form disulfide-linked heterodimers. It can dramatically accelerate the formation of intramolecular S-S bonds when added to an air-oxidized mixture of cysteine-containing peptides.
  • Useful reagent for sulfhydryl group determination
  • Common reagent in peptide chemistry
  • Used in oxidation-reduction condensations to form peptide bonds
  • Used in coupling reactions to form disulfide-linked heterodimers
  • Accelerates intramolecular S-S bond formation in cysteine-containing peptides