Learn More
Medchemexpress LLC 2,2'-Dipyridyl disulfide | 2127-03-9 | 99.9% | 220.32 | 5 G
SDP
Catalog No. 5000365970
Encompass_Preferred
2,2'-Dipyridyl disulfide is a chemical reagent primarily used for the determination of sulfhydryl groups. In peptide chemistry, this compound is valuable for its role in oxidation-reduction condensations, facilitating the creation of peptide bonds, and in coupling reactions to form disulfide-linked heterodimers.
  • Useful reagent for determining sulfhydryl groups
  • Employed in peptide chemistry for oxidation-reduction condensations
  • Facilitates creation of peptide bonds
  • Used in coupling reactions to form disulfide-linked heterodimers
  • Accelerates the formation of intramolecular S-S bonds
Catalog No. 50-003-65970 Supplier Medchemexpress LLC Supplier No. 5GHYY1666
2,2'-Dipyridyl disulfide is a chemical reagent primarily used for the determination of sulfhydryl groups. In peptide chemistry, this compound is valuable for its role in oxidation-reduction condensations, facilitating the creation of peptide bonds, and in coupling reactions to form disulfide-linked heterodimers.
  • Useful reagent for determining sulfhydryl groups
  • Employed in peptide chemistry for oxidation-reduction condensations
  • Facilitates creation of peptide bonds
  • Used in coupling reactions to form disulfide-linked heterodimers
  • Accelerates the formation of intramolecular S-S bonds