Learn More
Medchemexpress LLC 2,2'-Dipyridyl disulfide | 2127-03-9 | 99.93% | 220.32 | 10 G
SDP
Catalog No. 5000364958
Encompass_Preferred
2,2'-Dipyridyl disulfide is a useful reagent for the determination of sulfhydryl groups and a common reagent in peptide chemistry. It is often used in oxidation-reduction condensations to form peptide bonds or in coupling reactions to form disulfide-linked heterodimers. The formation of intramolecular S-S bonds can be dramatically accelerated by the addition of a 2,2'-Dipyridyl disulfide solution to an air-oxidized mixture of cysteine-containing peptides.
  • Useful reagent for sulfhydryl group determination
  • Common reagent in peptide chemistry
  • Used in oxidation-reduction condensations to form peptide bonds
  • Used in coupling reactions to form disulfide-linked heterodimers
  • Accelerates intramolecular S-S bond formation
Catalog No. 50-003-64958 Supplier Medchemexpress LLC Supplier No. 10GHYY1666
2,2'-Dipyridyl disulfide is a useful reagent for the determination of sulfhydryl groups and a common reagent in peptide chemistry. It is often used in oxidation-reduction condensations to form peptide bonds or in coupling reactions to form disulfide-linked heterodimers. The formation of intramolecular S-S bonds can be dramatically accelerated by the addition of a 2,2'-Dipyridyl disulfide solution to an air-oxidized mixture of cysteine-containing peptides.
  • Useful reagent for sulfhydryl group determination
  • Common reagent in peptide chemistry
  • Used in oxidation-reduction condensations to form peptide bonds
  • Used in coupling reactions to form disulfide-linked heterodimers
  • Accelerates intramolecular S-S bond formation