Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Dipyridyl disulfide, 98%

Catalog No. AAA1111814
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1111814 25 g
AAA1111806 5 g
AAA1111822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1111814 Supplier Thermo Scientific Chemicals Supplier No. A1111814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2127-03-9 | C10H8N2S2 | 220.31 g/mol

2,2'-Dipyridyl disulfide is a useful reagent for determination of sulfhydryl groups, preparation of amino acid active esters and the thio esters of phosphoric acid. It acts as a peptide coupling reagent and as an oxidizing agent. It is also used for the activation of glycosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Dipyridyl disulfide is a useful reagent for determination of sulfhydryl groups, preparation of amino acid active esters and the thio esters of phosphoric acid. It acts as a peptide coupling reagent and as an oxidizing agent. It is also used for the activation of glycosides.

Solubility
Soluble in water, benzene, tetrahydrofuran, dichloromethane, acetonitrile and dimethyformamide.

Notes
Incompatible with oxidizing agents and metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2127-03-9
Molecular Formula C10H8N2S2
Molecular Weight (g/mol) 220.31
MDL Number MFCD00006287
InChI Key HAXFWIACAGNFHA-UHFFFAOYSA-N
Synonym 2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide
PubChem CID 65093
IUPAC Name 2-(pyridin-2-yldisulfanyl)pyridine
SMILES C1=CC=NC(=C1)SSC2=CC=CC=N2

Specifications

Melting Point 56°C to 59°C
Flash Point 210°C (410°F)
Assay Percent Range 98%
Quantity 25 g
Beilstein 154629
Solubility Information Soluble in water,benzene,tetrahydrofuran,dichloromethane,acetonitrile and dimethyformamide.
Formula Weight 220.32
Percent Purity 98%
Chemical Name or Material 2,2'-Dipyridyl disulfide
missing translation for 'healthHazard1' H302-H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 218-343-1
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.