Learn More
Medchemexpress LLC 2 2 - Ethane-1 2-diy 5g
SDP
Catalog No. 5000205238
Encompass_Preferred
2 2 -(Ethane-1 2-diyl)dianiline (2 2 -Ethylenedianiline) can be utilized as precursor of metal complex[1]
Catalog No. 50-002-05238 Supplier Medchemexpress LLC Supplier No. HYW1040125G
2 2 -(Ethane-1 2-diyl)dianiline (2 2 -Ethylenedianiline) can be utilized as precursor of metal complex[1]