Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Ethoxyethoxy)ethyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. E065225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E065225G 25 g
E0652500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E065225G Supplier TCI America Supplier No. E065225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7328-17-8
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
InChI Key FTALTLPZDVFJSS-UHFFFAOYSA-N
Synonym Acrylic Acid 2-(2-Ethoxyethoxy)ethyl Ester, Diethylene Glycol Monoethyl Ether Acrylate
PubChem CID 81766
IUPAC Name 2-(2-ethoxyethoxy)ethyl prop-2-enoate
SMILES CCOCCOCCOC(=O)C=C

Specifications

Color Colorless
Boiling Point 250°C
Quantity 25 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Ethoxyethoxy)ethyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-3106
RTECSNumber UD3343500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.