Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2'-(Ethylenedioxy)bis(ethylamine)

Catalog No. 385506500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-550-6500ML 500 mL
38-550-6100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-550-6500ML Supplier Sigma Aldrich Supplier No. 385506500ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2G-(ethylenedioxy)-bis-(ethylamine) (EDEA) was used to linker to form conjugates of:manganese-doped zinc sulfide nanoparticles with folic acid and dimeric neomycin-neomycin. Glycidyl groups of the l-GMA particles were reacted with EDEA to produce large amine-functionalized particles (l-NH2). Polycondensation of guanidine hydrochloride and 2,2G-(ethylenedioxy)bis(ethylamine) has been used to measure biocides released into the environmental water samples.

Specifications

CAS 929-59-9
Density 1.015 g/mL (at 25°C)
Boiling Point 105°C to 109°C (6 mmHg)
Molecular Formula C6H16N2O2
Refractive Index n20/D 1.461 (literature)
Linear Formula NH2CH2CH2OCH2CH2OCH2CH2NH2
Quantity 500 mL
MDL Number MFCD00040474
Synonym 1,2-Bis(2-aminoethoxy)ethane; 1,8-Diamino-3,6-dioxaoctane; 2,2 -(Ethylenedioxy)diethylamine
Molecular Weight (g/mol) 148.2
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.