Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2'-(Ethylenedioxy)diethanethiol

Catalog No. 465178100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-517-8100ML 100 mL
46-517-8500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-517-8100ML Supplier Sigma Aldrich Supplier No. 465178100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,6-dioxa-1,8-octanedithiol is non volatile in nature.EDT can be crosslinked with unsaturated olive oil to form a photoinduced eco-friendly film through renewable sources. It can be used in the synthesis of bioanode for the fabrication of a biofuel cell (BFC) for glucose sensing application. It may also be used for other applications such as improving the optical properties of gold nanoclusters (AuNCs) and the development of bio-mediated synthetic polyester.

Specifications

CAS 14970-87-7
Density 1.12 g/mL (at 25°C)
Boiling Point 225°C
Molecular Formula C6H14O2S2
Refractive Index n20/D 1.509 (literature)
Linear Formula HSCH2CH2OCH2CH2OCH2CH2SH
Quantity 100 mL
MDL Number MFCD00015873
Synonym 1,2-Bis(2-mercaptoethoxy)ethane; 3,6-Dioxa-1,8-octane-dithiol
Molecular Weight (g/mol) 182.3
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.