Learn More
Medchemexpress LLC 2-[2-fluoro-4-[(2S)-2-pyrrolidinyl]phenyl]-1H-benzimidazole-7-carboxamide | 934162-61-5 | 99.0% | 324.35 g·mol⁻¹ | C18H17FN4O | 1 MG
SDP
Catalog No. 5000243599
Encompass_Preferred
A-966492 is a potent small-molecule inhibitor of poly(ADP-ribose) polymerase 1 (PARP1) and PARP2 used in preclinical research. It shows biochemical potency with Ki ≈ 1 nM for PARP1 and ≈ 1.5 nM for PARP2. Supplied as a high-purity solid suitable for in vitro and in vivo studies of DNA repair and oncology.
  • High biochemical potency: Ki ≈ 1 nM (PARP1) and ≈ 1.5 nM (PARP2).
  • High purity: 99.0% ensuring reproducible results.
  • Small-molecule solid suitable for dosing and formulation studies.
  • Stable under recommended storage conditions for long-term use.
  • Applicable to in vitro and in vivo studies of DNA repair and cancer biology.
Catalog No. 50-002-43599 Supplier Medchemexpress LLC Supplier No. HY106141MG
A-966492 is a potent small-molecule inhibitor of poly(ADP-ribose) polymerase 1 (PARP1) and PARP2 used in preclinical research. It shows biochemical potency with Ki ≈ 1 nM for PARP1 and ≈ 1.5 nM for PARP2. Supplied as a high-purity solid suitable for in vitro and in vivo studies of DNA repair and oncology.
  • High biochemical potency: Ki ≈ 1 nM (PARP1) and ≈ 1.5 nM (PARP2).
  • High purity: 99.0% ensuring reproducible results.
  • Small-molecule solid suitable for dosing and formulation studies.
  • Stable under recommended storage conditions for long-term use.
  • Applicable to in vitro and in vivo studies of DNA repair and cancer biology.