Learn More
Medchemexpress LLC 2-(2-fluorophenyl)-N-hydroxy-2-[4-[5-(trifluoromethyl)pyrimidin-2-yl]phenyl]acetamide | 1629729-98-1 | 98.1% | 50 MG
SDP
Catalog No. 5000234654
Encompass_Preferred
CHDI-390576 is a potent, cell-permeable, CNS-penetrant small-molecule inhibitor selective for class IIa histone deacetylases (HDAC4, HDAC5, HDAC7, HDAC9). It is provided as a high-purity research compound for biochemical and cellular studies of HDAC function and selectivity.
  • Low-nanomolar inhibitory activity against class IIa HDAC isoforms (reported IC50s: HDAC4 54 nM, HDAC5 60 nM, HDAC7 31 nM, HDAC9 50 nM).
  • High selectivity against class I HDACs and over HDAC8 and HDAC6.
  • CNS penetrant and cell permeable for central nervous system research.
  • High purity suitable for biochemical and cellular assays.
  • Molecular formula C19H13F4N3O2; molecular weight 391.32.
Catalog No. 50-002-34654 Supplier Medchemexpress LLC Supplier No. HY11993950MG
CHDI-390576 is a potent, cell-permeable, CNS-penetrant small-molecule inhibitor selective for class IIa histone deacetylases (HDAC4, HDAC5, HDAC7, HDAC9). It is provided as a high-purity research compound for biochemical and cellular studies of HDAC function and selectivity.
  • Low-nanomolar inhibitory activity against class IIa HDAC isoforms (reported IC50s: HDAC4 54 nM, HDAC5 60 nM, HDAC7 31 nM, HDAC9 50 nM).
  • High selectivity against class I HDACs and over HDAC8 and HDAC6.
  • CNS penetrant and cell permeable for central nervous system research.
  • High purity suitable for biochemical and cellular assays.
  • Molecular formula C19H13F4N3O2; molecular weight 391.32.