Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Hydroxy-5-methylphenyl)benzotriazole 99.0+%, TCI America™
SDP

Catalog No. H056025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H056025G 25 g
H0560500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H056025G Supplier TCI America Supplier No. H056025G
Add to Cart
Add to Cart

2-(2-Hydroxy-5-methylphenyl)benzotriazole, >-99.0% (HPLC,T), C13H11N3O, CAS Number-2440-22-4

2-(2-Hydroxy-5-methylphenyl)benzotriazole, >-99.0% (HPLC,T), C13H11N3O, CAS Number-2440-22-4

Chemical Identifiers

CAS 2440-22-4
Molecular Formula C13H11N3O
Molecular Weight (g/mol) 225.251
MDL Number MFCD00022903
InChI Key MCPKSFINULVDNX-UHFFFAOYSA-N
Synonym drometrizole, benazol p, tinuvin p, 2-2-benzotriazolyl-p-cresol, uv absorber-1, 2-2-hydroxy-5-methylphenyl benzotriazole, 2-2h-benzotriazol-2-yl-p-cresol, 2-2h-benzo d 1,2,3 triazol-2-yl-4-methylphenol, tin p, porex p
PubChem CID 17113
IUPAC Name 2-(benzotriazol-2-yl)-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)N2N=C3C=CC=CC3=N2

Specifications

Melting Point 130°C
Color Yellow
Quantity 25 g
Formula Weight 225.25
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Hydroxy-5-methylphenyl)benzotriazole
EINECSNumber (5)-0544
RTECSNumber GO6860000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.