Learn More
Medchemexpress LLC 2-(2-hydroxyphenyl)-1H-benzimidazole | 2963-66-8 | MFCD00022668 | 99.9% | 210.23 g·mol^-1 | C13H10N2O | 500 MG
SDP
Catalog No. 5000283603
Encompass_Preferred
2-(2′-Hydroxyphenyl)benzimidazole is a small-molecule fluorescent probe supplied as a solid research reagent. It is used in fluorescence spectroscopy, probe development, and analytical assays that require a reliable fluorescent dye.
  • High purity suitable for analytical use (99.9%).
  • Solid form for easy weighing and storage.
  • Well-defined molecular composition: C13H10N2O, MW 210.23 g·mol^-1.
  • Compatible with fluorescence spectroscopy and probe development.
  • Available in compact research package sizes such as 500 MG.
Catalog No. 50-002-83603 Supplier Medchemexpress LLC Supplier No. HYW140203500MG
2-(2′-Hydroxyphenyl)benzimidazole is a small-molecule fluorescent probe supplied as a solid research reagent. It is used in fluorescence spectroscopy, probe development, and analytical assays that require a reliable fluorescent dye.
  • High purity suitable for analytical use (99.9%).
  • Solid form for easy weighing and storage.
  • Well-defined molecular composition: C13H10N2O, MW 210.23 g·mol^-1.
  • Compatible with fluorescence spectroscopy and probe development.
  • Available in compact research package sizes such as 500 MG.