Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(2-hydroxyphenyl)benzimidazole | 2963-66-8 | MFCD00022668 | 99.9% | 210.23 g/mol | C13H10N2O | 1 ML
SDP

Catalog No. 5000283450
Encompass_Preferred

2-(2′-Hydroxyphenyl)benzimidazole is a fluorescent benzimidazole derivative provided as a pre-dissolved 10 mM solution in DMSO (1 mL). It is intended for laboratory research applications as a fluorescent probe. Use is restricted to research purposes and not for human or clinical use.

  • Supplied as a 10 mM solution in DMSO.
  • High purity (>99.9%).
  • CAS number 2963-66-8.
  • Molecular weight 210.23 g/mol.
  • Provided in a 1 mL vial convenient for small-scale assays.
  • Suitable as a fluorescent probe in laboratory research applications.

Catalog No. 50-002-83450 Supplier Medchemexpress LLC Supplier No. HYW14020310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(2′-Hydroxyphenyl)benzimidazole is a fluorescent benzimidazole derivative provided as a pre-dissolved 10 mM solution in DMSO (1 mL). It is intended for laboratory research applications as a fluorescent probe. Use is restricted to research purposes and not for human or clinical use.

  • Supplied as a 10 mM solution in DMSO.
  • High purity (>99.9%).
  • CAS number 2963-66-8.
  • Molecular weight 210.23 g/mol.
  • Provided in a 1 mL vial convenient for small-scale assays.
  • Suitable as a fluorescent probe in laboratory research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.