Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(2-Hydroxyphenyl)-N-(2-propen-1-yloxy)-1H-benzimidazole-6-carboxamide | 2404756-81-4 | 98.1% | 309.32 | C17H15N3O3 | 5 MG
SDP

Catalog No. 5000222527
Encompass_Preferred

X5050 is a small-molecule inhibitor of REST (RE1-silencing transcription factor) intended for research use. It inhibits REST with an EC50 of 2.1 μM and is provided as a solid with high purity for use in biochemical and cell-based assays. Recommended handling includes storage of powder at low temperatures and availability as pre-made DMSO solutions for convenience.

  • Inhibits REST with EC50 of 2.1 μM.
  • High purity: 98.1%.
  • Molecular weight 309.32 and formula C17H15N3O3.
  • Available as solid and as 10 mM solution in DMSO.
  • Appearance: white to light yellow solid.
  • Storage: powder at -20°C; solution at -80°C for long term.
  • Intended for research use only, not for human use.

Catalog No. 50-002-22527 Supplier Medchemexpress LLC Supplier No. HY1368335MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

X5050 is a small-molecule inhibitor of REST (RE1-silencing transcription factor) intended for research use. It inhibits REST with an EC50 of 2.1 μM and is provided as a solid with high purity for use in biochemical and cell-based assays. Recommended handling includes storage of powder at low temperatures and availability as pre-made DMSO solutions for convenience.

  • Inhibits REST with EC50 of 2.1 μM.
  • High purity: 98.1%.
  • Molecular weight 309.32 and formula C17H15N3O3.
  • Available as solid and as 10 mM solution in DMSO.
  • Appearance: white to light yellow solid.
  • Storage: powder at -20°C; solution at -80°C for long term.
  • Intended for research use only, not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.