Learn More
Medchemexpress LLC 2-(2-methoxyethoxy)ethanamine | 31576-51-9 | MFCD09032941 | 99.9% | 119.16 g/mol | C5H13NO2 | 1 G
SDP
Catalog No. 5000203184
Encompass_Preferred
m-PEG2-Amine is a methoxy-terminated polyethylene glycol (PEG2) linker bearing a terminal primary amine used for bioconjugation and medicinal chemistry. It is commonly employed as a PEG-based PROTAC linker and as a cleavable linker in antibody-drug conjugate (ADC) synthesis.
  • Methoxy-terminated PEG2 spacer with a terminal primary amine.
  • Suitable for PROTAC synthesis and as a cleavable ADC linker.
  • Hydrophilic spacer improves solubility and flexibility of conjugates.
  • High purity for reliable conjugation reactions.
  • Store at 4°C and protect from light; in solution, store long-term at -80°C.
Catalog No. 50-002-03184 Supplier Medchemexpress LLC Supplier No. HYW0084291G
m-PEG2-Amine is a methoxy-terminated polyethylene glycol (PEG2) linker bearing a terminal primary amine used for bioconjugation and medicinal chemistry. It is commonly employed as a PEG-based PROTAC linker and as a cleavable linker in antibody-drug conjugate (ADC) synthesis.
  • Methoxy-terminated PEG2 spacer with a terminal primary amine.
  • Suitable for PROTAC synthesis and as a cleavable ADC linker.
  • Hydrophilic spacer improves solubility and flexibility of conjugates.
  • High purity for reliable conjugation reactions.
  • Store at 4°C and protect from light; in solution, store long-term at -80°C.