Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] 98.0+%, TCI America™
SDP

Catalog No. M1402100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1402100MG 100 mg
M1402500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1402100MG Supplier TCI America Supplier No. M1402100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 175166-49-1
Molecular Formula C21H18N2O2
Molecular Weight (g/mol) 330.387
MDL Number MFCD00799561
InChI Key BDHSVQLSNIGJNC-JYWFKMLOSA-N
PubChem CID 11024010
IUPAC Name (3aR,8bS)-2-[[(3aR,8bS)-4,8b-dihydro-3aH-indeno[1,2-d][1,3]oxazol-2-yl]methyl]-4,8b-dihydro-3aH-indeno[1,2-d][1,3]oxazole
SMILES C1C2C(C3=CC=CC=C31)N=C(O2)CC4=NC5C(O4)CC6=CC=CC=C56

Specifications

Melting Point 224°C
Color White
Quantity 100 mg
Formula Weight 330.39
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (-)-2,2′-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.