Learn More
Medchemexpress LLC 2,2'-Methylenebis(4-methyl-6-tert-butylphenol) | 119-47-1 | 99.8% | 1 ML
SDP
Catalog No. 5000231055
Encompass_Preferred
This compound (CAS 119-47-1) is an acyl-CoA:cholesterol acyltransferase (ACAT) inhibitor isolated from Aspergillus fumigatus F93. It is supplied as a 10 mM solution in DMSO (1 mL), has molecular formula C23H32O2, molecular weight 340.50 g/mol, and a reported purity of 99.77%.
  • ACAT inhibitor useful for cancer research.
  • Supplied as a 10 mM solution in DMSO, ready for experimental use.
  • High reported purity suitable for biochemical assays.
  • Soluble in DMSO at 100 mg/mL (≈293.69 mM).
  • Stable in solvent when stored at -80°C (up to 2 years) or -20°C (up to 1 year).
Catalog No. 50-002-31055 Supplier Medchemexpress LLC Supplier No. HY12125410MM 1ML
This compound (CAS 119-47-1) is an acyl-CoA:cholesterol acyltransferase (ACAT) inhibitor isolated from Aspergillus fumigatus F93. It is supplied as a 10 mM solution in DMSO (1 mL), has molecular formula C23H32O2, molecular weight 340.50 g/mol, and a reported purity of 99.77%.
  • ACAT inhibitor useful for cancer research.
  • Supplied as a 10 mM solution in DMSO, ready for experimental use.
  • High reported purity suitable for biochemical assays.
  • Soluble in DMSO at 100 mg/mL (≈293.69 mM).
  • Stable in solvent when stored at -80°C (up to 2 years) or -20°C (up to 1 year).