Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,2'-methylenebis(4-methyl-6-tert-butylphenol) | 119-47-1 | 99.8% | 1000 G
SDP

Catalog No. 5000233478
Encompass_Preferred

This ACAT inhibitor (CAS 119-47-1) is supplied as a high-purity research-grade solid intended for biochemical and cancer research. Molecular formula C23H32O2, molecular weight 340.50 g/mol, purity 99.77%.

  • Inhibits acyl-CoA:cholesterol acyltransferase (ACAT).
  • Reported source: Aspergillus fumigatus F93.
  • Molecular formula C23H32O2 and molecular weight 340.50 g/mol.
  • High purity (99.77%).
  • Supplied as a solid in a 1000 G pack.

Catalog No. 50-002-33478 Supplier Medchemexpress LLC Supplier No. HY1212541000G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This ACAT inhibitor (CAS 119-47-1) is supplied as a high-purity research-grade solid intended for biochemical and cancer research. Molecular formula C23H32O2, molecular weight 340.50 g/mol, purity 99.77%.

  • Inhibits acyl-CoA:cholesterol acyltransferase (ACAT).
  • Reported source: Aspergillus fumigatus F93.
  • Molecular formula C23H32O2 and molecular weight 340.50 g/mol.
  • High purity (99.77%).
  • Supplied as a solid in a 1000 G pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.