Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,2'-methylenebis(4-methyl-6-tert-butylphenol) | 119-47-1 | 99.8% | 250 G
SDP

Catalog No. 5000231407
Encompass_Preferred

GERI-BP002-A (CAS 119-47-1) is a small-molecule inhibitor of acyl-CoA:cholesterol acyltransferase (ACAT) isolated from Aspergillus fumigatus F93 and supplied as a high-purity solid for laboratory research.

  • High purity: 99.77%.
  • Molecular formula C23H32O2; molecular weight 340.50 g/mol.
  • Solid form; white to off-white appearance.
  • Soluble in DMSO; formulation protocols provided for in vivo use.
  • Stable under recommended storage conditions; in solution, -80°C (up to 2 years) and -20°C (up to 1 year).
  • Intended for research use only; not for diagnostic or therapeutic use.

Catalog No. 50-002-31407 Supplier Medchemexpress LLC Supplier No. HY121254250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GERI-BP002-A (CAS 119-47-1) is a small-molecule inhibitor of acyl-CoA:cholesterol acyltransferase (ACAT) isolated from Aspergillus fumigatus F93 and supplied as a high-purity solid for laboratory research.

  • High purity: 99.77%.
  • Molecular formula C23H32O2; molecular weight 340.50 g/mol.
  • Solid form; white to off-white appearance.
  • Soluble in DMSO; formulation protocols provided for in vivo use.
  • Stable under recommended storage conditions; in solution, -80°C (up to 2 years) and -20°C (up to 1 year).
  • Intended for research use only; not for diagnostic or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.