Learn More
Medchemexpress LLC 2,2'-methylenebis(4-methyl-6-tert-butylphenol) | 119-47-1 | 99.8% | 500 G

Supplier: Medchemexpress LLC HY121254500G
GERI-BP002-A is a small-molecule inhibitor of acyl-CoA:cholesterol acyltransferase (ACAT) isolated from Aspergillus fumigatus F93, provided for research use with reported cytotoxic activity against multiple cancer cell lines.
- High purity (99.8%) for reliable experimental results.
- Molecular formula C23H32O2 and molecular weight 340.50 g·mol⁻¹.
- Soluble in DMSO (100 mg/mL); ultrasonic assistance recommended.
- Storage stability in solvent: -80°C (2 years), -20°C (1 year).
- Available in multiple pack sizes up to 500 G for scale flexibility.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.