Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Naphthyl)indole 98.0+%, TCI America™
SDP

Catalog No. N08591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08591G Supplier TCI America Supplier No. N08591G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 23746-81-8
Molecular Formula C18H13N
Molecular Weight (g/mol) 243.31
MDL Number MFCD00124144
InChI Key CACDYUNTCMPDMI-UHFFFAOYSA-N
Synonym 2-2-naphthyl indole, 2-2-naphthyl-1h-indole, 2-naphthalen-2-yl-1h-indole, acmc-1chvp, 2-2-naphthyl-indol, maybridge1_005904, 2-2-indolyl naphthalene, 2-1h-indol-2-yl naphthalene, 2-2-naphthalenyl-1h-indole
PubChem CID 265232
IUPAC Name 2-(naphthalen-2-yl)-1H-indole
SMILES N1C2=CC=CC=C2C=C1C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 204°C
Quantity 1 g
Formula Weight 243.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Naphthyl)indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.