Learn More
Medchemexpress LLC 2,2-Oxybis(ethylamine) | 2752-17-2 | 98.0% | C4H12N2O | 500 MG
SDP
Catalog No. 5000356728
Encompass_Preferred
2,2-Oxybis(ethylamine) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-56728 Supplier Medchemexpress LLC Supplier No. HY140208500MG
2,2-Oxybis(ethylamine) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only