Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[2,2]-Paracyclophane, 99%

Catalog No. p-7077221
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC291600010 1 g
AC291600050 5 g
AC291600250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC291600010 Supplier Thermo Scientific Chemicals Supplier No. 291600010
Only null left
Add to Cart

CAS: 1633-22-3 | C16H16 | 208.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1633-22-3
Molecular Formula C16H16
Molecular Weight (g/mol) 208.3
InChI Key OOLUVSIJOMLOCB-UHFFFAOYSA-N
Synonym 2.2 paracyclophane, di-p-xylylene, di-1,4-xylylene, paracyclophane, cyclobis benzene-1,4-dimethylene, tricyclo 8,2,2,24,7 hexadeca-4,6,10,12,13,15-hexaene, 2.2-paracyclophane, tricyclo 8.2.2.24,7 hexadeca-4,6,10,12,13,15-hexaene, 2,2-paracyclophane
PubChem CID 74210
SMILES C1CC2=CC=C(CCC3=CC=C1C=C3)C=C2

Specifications

Melting Point 285.0°C to 288.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene
Formula Weight 208.3
Percent Purity 99%
Chemical Name or Material [2,2]-Paracyclophane
EINECSNumber 216-644-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.