Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[(2-Phenoxyethyl)thio]acetic acid, 97%, Thermo Scientific™

Catalog No. KM06649DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KM06649DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM06649DA Supplier Thermo Scientific Chemicals Supplier No. KM06649DA
Add to Cart

Chemical Identifiers

CAS 75434-70-7
Molecular Formula C10H12O3S
Molecular Weight (g/mol) 212.263
MDL Number MFCD00085152
InChI Key SHWKIHWUFNTMIB-UHFFFAOYSA-N
Synonym 2-2-phenoxyethyl thio acetic acid, 2-2-phenoxyethyl sulfanyl acetic acid, 2-phenoxyethyl sulfanyl acetic acid, maybridge1_005030, 2-2-phenoxyethylthio acetic acid, 2-phenoxy-ethylsulfanyl acetic acid, 2-phenoxy-ethylsulfanyl-acetic acid, 2-2-phenoxyethylsulfanyl acetic acid
PubChem CID 2779087
IUPAC Name 2-(2-phenoxyethylsulfanyl)acetic acid
SMILES C1=CC=C(C=C1)OCCSCC(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula C6H5O(CH2)2SCH2CO2H
Quantity 1 g
Formula Weight 212.27
Percent Purity 97%
Chemical Name or Material 2-[(2-Phenoxyethyl)thio]acetic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.