Learn More
Medchemexpress LLC 2-(2-Phenylethyl)chromone | 61828-53-3 | 50 MG
SDP
Catalog No. 5000417389
Encompass_Preferred
2-(2-Phenylethyl)chromone (Flidersiachromone) is a chromone derivative and antioxidant. It scavenges ABTS·+ free radicals, and its derivatives show various activities, including anti-inflammatory, neuroprotective, anti-tumor, and α-glucosidase inhibitory effects. This product is suitable for experimental research in areas such as inflammation-related diseases, tumors, and neurodegenerative diseases.
  • Chromone derivative and antioxidant
  • Scavenges ABTS·+ free radicals
  • Derivatives exhibit diverse activities, such as anti-inflammatory, neuroprotective, anti-tumor, and α-glucosidase inhibitory effects
  • Applicable for experimental research in inflammation-related diseases, tumors, and neurodegenerative diseases
  • Purity: 99.94%
  • Derivatives show α-glucosidase inhibitory activity
  • Derivatives demonstrate neuroprotective activity by protecting human neuroblastoma SH-SY5Y cells against H2O2-induced apoptosis
Catalog No. 50-004-17389 Supplier Medchemexpress LLC Supplier No. HYN822050MG
2-(2-Phenylethyl)chromone (Flidersiachromone) is a chromone derivative and antioxidant. It scavenges ABTS·+ free radicals, and its derivatives show various activities, including anti-inflammatory, neuroprotective, anti-tumor, and α-glucosidase inhibitory effects. This product is suitable for experimental research in areas such as inflammation-related diseases, tumors, and neurodegenerative diseases.
  • Chromone derivative and antioxidant
  • Scavenges ABTS·+ free radicals
  • Derivatives exhibit diverse activities, such as anti-inflammatory, neuroprotective, anti-tumor, and α-glucosidase inhibitory effects
  • Applicable for experimental research in inflammation-related diseases, tumors, and neurodegenerative diseases
  • Purity: 99.94%
  • Derivatives show α-glucosidase inhibitory activity
  • Derivatives demonstrate neuroprotective activity by protecting human neuroblastoma SH-SY5Y cells against H2O2-induced apoptosis