Learn More
Medchemexpress LLC 2-(2-Phenylethyl)chromone (Flidersiachromone) | 61828-53-3 | 100 MG
SDP
Catalog No. 5000419698
Encompass_Preferred
2-(2-Phenylethyl)chromone (Flidersiachromone) is a chromone derivative that functions as an antioxidant. It is capable of scavenging ABTS·+ free radicals. Derivatives of this compound have demonstrated a range of activities, including anti-inflammatory, neuroprotective, anti-tumor, and α-glucosidase inhibitory effects. It is suitable for experimental research in areas such as inflammation-related diseases, tumors, and neurodegenerative diseases.
  • Chromone derivative.
  • Acts as an antioxidant agent.
  • Scavenges ABTS·+ free radicals.
  • Derivatives exhibit anti-inflammatory effects.
  • Derivatives exhibit neuroprotective effects.
  • Derivatives exhibit anti-tumor effects.
  • Derivatives show α-glucosidase inhibitory effects.
  • Applicable to research in inflammation-related diseases.
  • Applicable to research in tumors.
  • Applicable to research in neurodegenerative diseases.
Catalog No. 50-004-19698 Supplier Medchemexpress LLC Supplier No. HYN8220100MG
2-(2-Phenylethyl)chromone (Flidersiachromone) is a chromone derivative that functions as an antioxidant. It is capable of scavenging ABTS·+ free radicals. Derivatives of this compound have demonstrated a range of activities, including anti-inflammatory, neuroprotective, anti-tumor, and α-glucosidase inhibitory effects. It is suitable for experimental research in areas such as inflammation-related diseases, tumors, and neurodegenerative diseases.
  • Chromone derivative.
  • Acts as an antioxidant agent.
  • Scavenges ABTS·+ free radicals.
  • Derivatives exhibit anti-inflammatory effects.
  • Derivatives exhibit neuroprotective effects.
  • Derivatives exhibit anti-tumor effects.
  • Derivatives show α-glucosidase inhibitory effects.
  • Applicable to research in inflammation-related diseases.
  • Applicable to research in tumors.
  • Applicable to research in neurodegenerative diseases.