Learn More
Medchemexpress LLC 2-[(2-phenylethyl)thio]nicotinic acid | 496775-95-2 | MFCD03621206 | 99.8% | 259.32 g/mol | C14H13NO2S | 10MG
SDP
Catalog No. 5000237749
Encompass_Preferred
ML-099 (2-[(2-phenylethyl)thio]nicotinic acid) is a small-molecule pan Ras-family GTPase activator used in biochemical and cell-based research to activate Rac1, CDC42, Ras, Rab7, and Rab-2A. It is supplied for research use and is characterized by high purity and a molecular weight of 259.32 g/mol.
  • Activates multiple Ras-family GTPases including Rac1 and CDC42.
  • Useful for cell-based and biochemical assays probing GTPase signaling.
  • High purity for research applications (reported 99.75%).
  • Molecular formula C14H13NO2S and molecular weight 259.32 g/mol.
  • Identified by CAS 496775-95-2 and PubChem CID 888706.
Catalog No. 50-002-37749 Supplier Medchemexpress LLC Supplier No. HY12430610MG
ML-099 (2-[(2-phenylethyl)thio]nicotinic acid) is a small-molecule pan Ras-family GTPase activator used in biochemical and cell-based research to activate Rac1, CDC42, Ras, Rab7, and Rab-2A. It is supplied for research use and is characterized by high purity and a molecular weight of 259.32 g/mol.
  • Activates multiple Ras-family GTPases including Rac1 and CDC42.
  • Useful for cell-based and biochemical assays probing GTPase signaling.
  • High purity for research applications (reported 99.75%).
  • Molecular formula C14H13NO2S and molecular weight 259.32 g/mol.
  • Identified by CAS 496775-95-2 and PubChem CID 888706.