Learn More
Medchemexpress LLC 2-[(2-phenylethyl)thio]nicotinic acid | 496775-95-2 | MFCD03621206 | 99.8% | 259.32 g/mol | C14H13NO2S | 50MG
SDP
Catalog No. 5000235232
Encompass_Preferred
ML-099 (CID-888706) is a small-molecule pan activator of Ras-related GTPases provided for research use. It enhances GTPase activity by increasing affinity for guanine nucleotides and activates multiple small GTPases relevant to cytoskeletal dynamics and signaling pathways. The compound is supplied as a solid in several pack sizes and as a DMSO solution for cell-based and biochemical assays.
Catalog No. 50-002-35232 Supplier Medchemexpress LLC Supplier No. HY12430650MG
ML-099 (CID-888706) is a small-molecule pan activator of Ras-related GTPases provided for research use. It enhances GTPase activity by increasing affinity for guanine nucleotides and activates multiple small GTPases relevant to cytoskeletal dynamics and signaling pathways. The compound is supplied as a solid in several pack sizes and as a DMSO solution for cell-based and biochemical assays.