Learn More
Medchemexpress LLC 2-[(2-phenylethyl)thio]nicotinic acid | 496775-95-2 | MFCD03621206 | 99.8% | 259.33 g/mol | C14H13NO2S | 100MG
SDP
Catalog No. 5000236827
Encompass_Preferred
ML-099 (CID-888706) is a small-molecule pan activator of Ras-related GTPases that activates Rac1, Cdc42, Ras, Rab7, and Rab-2A. Supplied as a solid research reagent with high analytical purity, it is used to probe Rho-family GTPase signaling in biochemical and cellular assays.
Catalog No. 50-002-36827 Supplier Medchemexpress LLC Supplier No. HY124306100MG
ML-099 (CID-888706) is a small-molecule pan activator of Ras-related GTPases that activates Rac1, Cdc42, Ras, Rab7, and Rab-2A. Supplied as a solid research reagent with high analytical purity, it is used to probe Rho-family GTPase signaling in biochemical and cellular assays.