Learn More
Medchemexpress LLC 2-(2-phenylethylsulfanyl)pyridine-3-carboxylic acid | 496775-95-2 | MFCD03621206 | 99.8% | 259.32 g·mol⁻1 | C14H13NO2S | 25MG
SDP
Catalog No. 5000235141
Encompass_Preferred
ML-099 (CID-888706) is a small-molecule pan Ras-related GTPases activator used in biochemical and cell-based research to activate Rac1, Cdc42, Ras, Rab7, and Rab-2A. It is supplied for research use only and is provided in high-purity solid form or as a DMSO solution suitable for in vitro assays.
  • Activates multiple small GTPases including Rac1, Cdc42, Ras, Rab7, and Rab-2A.
  • High purity suitable for biochemical and cell-based assays (99.8%).
  • Available as solid in small milligram quantities and as a 10 mM solution in DMSO.
  • Molecular formula C14H13NO2S and molecular weight 259.32 g·mol⁻1.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-35141 Supplier Medchemexpress LLC Supplier No. HY12430625MG
Add to Cart
Add to Cart
ML-099 (CID-888706) is a small-molecule pan Ras-related GTPases activator used in biochemical and cell-based research to activate Rac1, Cdc42, Ras, Rab7, and Rab-2A. It is supplied for research use only and is provided in high-purity solid form or as a DMSO solution suitable for in vitro assays.
  • Activates multiple small GTPases including Rac1, Cdc42, Ras, Rab7, and Rab-2A.
  • High purity suitable for biochemical and cell-based assays (99.8%).
  • Available as solid in small milligram quantities and as a 10 mM solution in DMSO.
  • Molecular formula C14H13NO2S and molecular weight 259.32 g·mol⁻1.
  • Intended for research use only; not for human or clinical use.