Learn More
Medchemexpress LLC 2,2'-(Piperazine-1,4-diyl)diethanamine | 6531-38-0 | 172.27 | 100 MG
SDP
Catalog No. 5000448902
Encompass_Preferred
2,2'-(Piperazine-1,4-diyl)diethanamine is a piperazine with two N-ethanamine groups. This type of molecule may be used to build drug molecules or design novel ionizable aminolipid head groups for lipid nanoparticles. For research use only.
  • Utilized in the construction of drug molecules.
  • Designed for novel ionizable aminolipid head groups.
  • Applicable in lipid nanoparticle research.
  • Intended for research use only.
Catalog No. 50-004-48902 Supplier Medchemexpress LLC Supplier No. HYW004440100MG
2,2'-(Piperazine-1,4-diyl)diethanamine is a piperazine with two N-ethanamine groups. This type of molecule may be used to build drug molecules or design novel ionizable aminolipid head groups for lipid nanoparticles. For research use only.
  • Utilized in the construction of drug molecules.
  • Designed for novel ionizable aminolipid head groups.
  • Applicable in lipid nanoparticle research.
  • Intended for research use only.