Learn More
Medchemexpress LLC 2,2'-(Piperazine-1,4-diyl)diethanamine | 6531-38-0 | 172.27 | 250 MG
SDP
Catalog No. 5000449973
Encompass_Preferred
2,2'-(Piperazine-1,4-diyl)diethanamine is a piperazine with two N-ethanamine groups. This type of molecule may be used to build drug molecules or design novel ionizable aminolipid head groups for lipid nanoparticles.
  • Piperazine derivative
  • Suitable for building drug molecules
  • Can be used to design novel ionizable aminolipid head groups for lipid nanoparticles
  • White to light yellow solid appearance
  • For research use only
Catalog No. 50-004-49973 Supplier Medchemexpress LLC Supplier No. 250MGHYW004440
2,2'-(Piperazine-1,4-diyl)diethanamine is a piperazine with two N-ethanamine groups. This type of molecule may be used to build drug molecules or design novel ionizable aminolipid head groups for lipid nanoparticles.
  • Piperazine derivative
  • Suitable for building drug molecules
  • Can be used to design novel ionizable aminolipid head groups for lipid nanoparticles
  • White to light yellow solid appearance
  • For research use only