Learn More
Medchemexpress LLC 2,2'-(Piperazine-1,4-diyl)diethanamine | 6531-38-0 | 172.27 | 500 MG
SDP
Catalog No. 5000446270
Encompass_Preferred
2,2'-(Piperazine-1,4-diyl)diethanamine is a piperazine compound that contains two N-ethanamine groups. This molecule can be utilized as a building block for drug molecules or for designing new ionizable aminolipid head groups, which are components for lipid nanoparticles. The product is intended for research use only and is not sold to patients.
  • Classified as a biochemical assay reagent.
  • Can be used to construct drug molecules.
  • Suitable for designing novel ionizable aminolipid head groups for lipid nanoparticles.
Catalog No. 50-004-46270 Supplier Medchemexpress LLC Supplier No. HYW004440500MG
2,2'-(Piperazine-1,4-diyl)diethanamine is a piperazine compound that contains two N-ethanamine groups. This molecule can be utilized as a building block for drug molecules or for designing new ionizable aminolipid head groups, which are components for lipid nanoparticles. The product is intended for research use only and is not sold to patients.
  • Classified as a biochemical assay reagent.
  • Can be used to construct drug molecules.
  • Suitable for designing novel ionizable aminolipid head groups for lipid nanoparticles.