Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(2-pyridinyl)-4H-1,3-benzothiazin-4-one | 99420-15-2 | MFCD19690951 | 99.3% | 240.28 g/mol | C13H8N2OS | 5 MG
SDP

Catalog No. 5000263506
Encompass_Preferred

BTZO-1 is a small-molecule research probe that binds macrophage migration inhibitory factor (MIF) and activates antioxidant response element (ARE)-mediated gene expression. Supplied as a solid for biochemical and cell-based assays, the compound is accompanied by stability and solubility data to support experimental planning.

  • Binds MIF with Kd ≈ 68.6 nM.
  • High purity (≈99.3%) suitable for research applications.
  • Molecular weight 240.28 g/mol; formula C13H8N2OS.
  • Soluble in DMSO at 8.33 mg/mL (requires ultrasonic).
  • Recommended storage: powder -20°C for long-term stability.

Catalog No. 50-002-63506 Supplier Medchemexpress LLC Supplier No. HY1100845MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BTZO-1 is a small-molecule research probe that binds macrophage migration inhibitory factor (MIF) and activates antioxidant response element (ARE)-mediated gene expression. Supplied as a solid for biochemical and cell-based assays, the compound is accompanied by stability and solubility data to support experimental planning.

  • Binds MIF with Kd ≈ 68.6 nM.
  • High purity (≈99.3%) suitable for research applications.
  • Molecular weight 240.28 g/mol; formula C13H8N2OS.
  • Soluble in DMSO at 8.33 mg/mL (requires ultrasonic).
  • Recommended storage: powder -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.