Learn More
Medchemexpress LLC 2-(2′-Pyridyl)benzimidazole | 1137-68-4 | 99.88% | 195.22 | 50 G
SDP
Catalog No. 5000452484
Encompass_Preferred
2-(2′-Pyridyl)benzimidazole is a potentially tridentate ligand that can form stable complexes with various transition metal ions. It can be used as an anti-inflammatory agent and an inhibitor of *Escherichia coli* methionine aminopeptidase.
  • Potentially tridentate ligand that can form stable complexes with various transition metal ions.
  • Can be used as an anti-inflammatory agent.
  • Can be used as an inhibitor of *Escherichia coli* methionine aminopeptidase.
Catalog No. 50-004-52484 Supplier Medchemexpress LLC Supplier No. HYW00990650G
2-(2′-Pyridyl)benzimidazole is a potentially tridentate ligand that can form stable complexes with various transition metal ions. It can be used as an anti-inflammatory agent and an inhibitor of *Escherichia coli* methionine aminopeptidase.
  • Potentially tridentate ligand that can form stable complexes with various transition metal ions.
  • Can be used as an anti-inflammatory agent.
  • Can be used as an inhibitor of *Escherichia coli* methionine aminopeptidase.