Learn More
Medchemexpress LLC 2-(2'-Pyridyl)benzimidazole | 1137-68-4 | 99.9% | 195.22 | 25 G
SDP
Catalog No. 5000366439
Encompass_Preferred
2-(2'-Pyridyl)benzimidazole is a potentially tridentate ligand capable of forming stable complexes with various transition metal ions. It functions as an anti-inflammatory agent and an inhibitor of *Escherichia coli* methionine aminopeptidase.
  • Potentially tridentate ligand that can form stable complexes with various transition metal ions.
  • Can be used as an anti-inflammatory agent.
  • Functions as an inhibitor of *Escherichia coli* methionine aminopeptidase.
Catalog No. 50-003-66439 Supplier Medchemexpress LLC Supplier No. 25GHYW009906
2-(2'-Pyridyl)benzimidazole is a potentially tridentate ligand capable of forming stable complexes with various transition metal ions. It functions as an anti-inflammatory agent and an inhibitor of *Escherichia coli* methionine aminopeptidase.
  • Potentially tridentate ligand that can form stable complexes with various transition metal ions.
  • Can be used as an anti-inflammatory agent.
  • Functions as an inhibitor of *Escherichia coli* methionine aminopeptidase.