Learn More
Medchemexpress LLC 2-(2'-Pyridyl)benzimidazole | 1137-68-4 | 99.9% | 195.22 | 5 G
SDP
Catalog No. 5000453520
Encompass_Preferred
2-(2'-Pyridyl)benzimidazole is a potentially tridentate ligand capable of forming stable complexes with various transition metal ions. It can also function as an anti-inflammatory agent and an inhibitor of *Escherichia coli* methionine aminopeptidase.
  • Potentially tridentate ligand
  • Forms stable complexes with various transition metal ions
  • Can be used as an anti-inflammatory agent
  • Functions as an inhibitor of *Escherichia coli* methionine aminopeptidase
Catalog No. 50-004-53520 Supplier Medchemexpress LLC Supplier No. HYW0099065G
Add to Cart
Add to Cart
2-(2'-Pyridyl)benzimidazole is a potentially tridentate ligand capable of forming stable complexes with various transition metal ions. It can also function as an anti-inflammatory agent and an inhibitor of *Escherichia coli* methionine aminopeptidase.
  • Potentially tridentate ligand
  • Forms stable complexes with various transition metal ions
  • Can be used as an anti-inflammatory agent
  • Functions as an inhibitor of *Escherichia coli* methionine aminopeptidase