Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Pyridyl)indole 97.0+%, TCI America™
SDP

Catalog No. P2142200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2142200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2142200MG Supplier TCI America Supplier No. P2142200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13228-40-5
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.237
MDL Number MFCD00033466
InChI Key OLGGLCIDAMICTA-UHFFFAOYSA-N
PubChem CID 270302
IUPAC Name 2-pyridin-2-yl-1H-indole
SMILES C1=CC=C2C(=C1)C=C(N2)C3=CC=CC=N3

Specifications

Melting Point 154°C
Quantity 200 mg
Formula Weight 194.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Pyridyl)indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.