Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Thienyl)ethanol 99%, Thermo Scientific™

Catalog No. AC138661000
Change view
Click to view available options
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC138661000 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC138661000 Supplier Thermo Scientific Chemicals Supplier No. 138661000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5402-55-1
CAS Min % 98.5
CAS Max % 100.0
Density 1.15
Infrared Spectrum Authentic
Molecular Formula C6H8OS
Refractive Index 1.5490 to 1.5520 (20°C, 589nm)
MDL Number MFCD00005462
Quantity 100mL
Beilstein 17, V,3, 379
Synonym 2-thiopheneethanol, 2-2-thienyl ethanol, thiophene-2-ethanol, 2-thiophen-2-yl ethanol, 2-2-hydroxyethyl thiophene, 2-thiophen-2-yl ethan-1-ol, ethanol, 2-2-thienyl, 2-thiophenethanol, 2-2-thienyl-ethanol, 2-thienylethanol
InChI Key VMJOFTHFJMLIKL-UHFFFAOYSA-N
SMILES OCCC1=CC=CS1
IUPAC Name 2-(thiophen-2-yl)ethan-1-ol
Molecular Weight (g/mol) 128.19
PubChem CID 79400
Formula Weight 128.19
Percent Purity ≥98.5%
Chemical Name or Material 2-(2-Thienyl)ethanol, 99%
Show More Show Less
EINECSNumber 226-452-0
TSCA TSCA
Recommended Storage this product discolors on storage
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.