Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Thienyl)ethyl isocyanate, 95+%, Thermo Scientific™

Catalog No. RF05240DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RF05240DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RF05240DA Supplier Thermo Scientific Chemicals Supplier No. RF05240DA
Add to Cart

Chemical Identifiers

CAS 58749-51-2
Molecular Formula C7H7NOS
Molecular Weight (g/mol) 153.199
InChI Key KAJZMNLREUEZSH-UHFFFAOYSA-N
PubChem CID 2782179
IUPAC Name 2-(2-isocyanatoethyl)thiophene
SMILES C1=CSC(=C1)CCN=C=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-2-isocyanatoethyl thiophene, 2-2-thienyl ethyl isocyanate, 2-thien-2-yl ethyl isocyanate, thiophene,2-2-isocyanatoethyl, 2-2-isocyanato-ethyl-thiophene, 2-2-thienyl ethanisocyanate, 2-2-isocyanatoethyl-thiophene, thiophene, 2-2-isocyanatoethyl, 2-thien-2-yl ethylisocyanate
Formula Weight 153.2
Percent Purity 95%
Chemical Name or Material 2-(2-Thienyl)ethyl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.