Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Thiobis(4,6-dichlorophenol) Disodium Salt 98.0+%, TCI America™
SDP

Catalog No. T040625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T040625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T040625G Supplier TCI America Supplier No. T040625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6385-58-6
Molecular Formula C12H4Cl4Na2O2S
Molecular Weight (g/mol) 400.002
MDL Number MFCD00058975
InChI Key FNYZFZRGBBCWBI-UHFFFAOYSA-L
Synonym Bis(3,5-dichloro-2-hydroxyphenyl) Sulfide Disodium Salt, Bithionol Disodium Salt
PubChem CID 22882
IUPAC Name disodium;2,4-dichloro-6-(3,5-dichloro-2-oxidophenyl)sulfanylphenolate
SMILES C1=C(C=C(C(=C1Cl)[O-])SC2=CC(=CC(=C2[O-])Cl)Cl)Cl.[Na+].[Na+]

Specifications

Quantity 25 g
Formula Weight 400.00
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Thiobis(4,6-dichlorophenol) Disodium Salt
EINECSNumber (3)-2437
RTECSNumber SN0700000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.