Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2'-Thiodiethanethiol

Catalog No. M400710G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M400710G 10 g
M400750G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M400710G Supplier Sigma Aldrich Supplier No. M400710G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2'-Thiodiethanethiol

Specifications

CAS 3570-55-6
Density 1.183 g/mL (at 25°C (literature))
Boiling Point 135°C to 136°C (10 mmHg)
Molecular Formula C4H10S3
Refractive Index n20/D 1.5961 (literature)
Linear Formula (HSCH2CH2)2 S
MDL Number MFCD00004893
Quantity 10 g
Synonym 2-Mercaptoethyl sulfide; Bis(2-mercaptoethyl) sulfide
Molecular Weight (g/mol) 154.32
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.