Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2'-Thiodiethanethiol

Catalog No. M400750G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M400750G 50 g
M400710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M400750G Supplier Sigma Aldrich Supplier No. M400750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2'-Thiodiethanethiol

Specifications

CAS 3570-55-6
Density 1.183 g/mL (at 25°C (literature))
Boiling Point 135°C to 136°C (10 mmHg)
Molecular Formula C4H10S3
Linear Formula (HSCH2CH2)2 S
Refractive Index n20/D 1.5961 (literature)
MDL Number MFCD00004893
Quantity 50 g
Synonym 2-Mercaptoethyl sulfide; Bis(2-mercaptoethyl) sulfide
Molecular Weight (g/mol) 154.32
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.