Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Thiodiethanethiol, tech. 85%

Catalog No. p-7026346
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2230409 10 g
AAB2230418 50 g
AAB2230430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2230409 Supplier Thermo Scientific Chemicals Supplier No. B2230409
Only null left
Add to Cart

CAS: 3570-55-6 | C4H10S3 | 154.30 g/mol

2,2?-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2'-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2′-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3570-55-6
Molecular Formula C4H10S3
Molecular Weight (g/mol) 154.30
MDL Number MFCD00004893
InChI Key KSJBMDCFYZKAFH-UHFFFAOYSA-N
Synonym 2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi
PubChem CID 77117
SMILES SCCSCCS

Specifications

Density 1.183
Boiling Point 135°C to 136°C (10 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.5955
Quantity 10 g
Solubility Information Difficult to mix.
IUPAC Name 2-(2-sulfanylethylsulfanyl)ethanethiol
Formula Weight 154.32
Percent Purity ≥90%
Chemical Name or Material 2,2'-Thiodiethanethiol
Hazard Category H227
Hazard Statement GHS H Statement
H332-H315-H319-H335-H227
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 222-671-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.