Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[2-(Trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
SDP

Catalog No. T24285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24285G Supplier TCI America Supplier No. T24285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1073339-21-5
Molecular Formula C13H16BF3O2
Molecular Weight (g/mol) 272.074
MDL Number MFCD06795676
InChI Key HBQNDHPCMDZKNT-UHFFFAOYSA-N
Synonym 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(trifluoromethyl)benzene
PubChem CID 17750282
IUPAC Name 4,4,5,5-tetramethyl-2-[2-(trifluoromethyl)phenyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C(F)(F)F

Specifications

Color Yellow
Boiling Point 73°C
Quantity 5 g
Formula Weight 272.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-[2-(Trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.