Learn More
Electronic Imaging Materials Inc Microplate 667 CryoLabel 2.20 inch x 0.25 inch, White, 1 inch core, 9000/roll (3K sets of 3)
SDP
Catalog No. NC1211670
Encompass_Preferred
2.20 inch x 0.25 inch white CryoLabel for Microplates. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and a wide variety of other chemicals. Designed for thermal transfer and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.
Catalog No. NC1211670 Supplier Electronic Imaging Materials Inc Supplier No. 92268
2.20 inch x 0.25 inch white CryoLabel for Microplates. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and a wide variety of other chemicals. Designed for thermal transfer and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.