Learn More
AMPLYUS LLC miniPCR bio 2-200 µl micropipette tips – 2 racks of 96
SDP
Catalog No. NC3736332
Encompass_Preferred
These 2-200 µl micropipette tips are the perfect companion for any standard 2-20 µl or 20-200 µl micropipette. Universal fit, 2 racks of 96 tips each. Tips are sterile, autoclavable, and certified to be free of any detectable RNase, DNase, DNA, or other contaminants.
Catalog No. NC3736332 Supplier AMPLYUS LLC Supplier No. CM100101
These 2-200 µl micropipette tips are the perfect companion for any standard 2-20 µl or 20-200 µl micropipette. Universal fit, 2 racks of 96 tips each. Tips are sterile, autoclavable, and certified to be free of any detectable RNase, DNase, DNA, or other contaminants.