Learn More
CELLTREAT Scientific Products 2-20UL Adjustable Pipette, Single Channel
SDP
Catalog No. NC1772264
Encompass_Preferred
CELLTREAT Scientific Products 2-20uL Adjustable Pipette is a high precision pipette with easy-to-read volume display. Continuously adjustable with locking rig to prevent volume changes while pipetting. Lightweight design and easy operation with either hand. Reduced operating forces: plunger is easy to press and dispense. Stainless steel and PTFE components for long life. It is not receommended to autocalve more than 6 times. Fits CELLTREAT Scientific Products Universal Pipette Tips for accurate pipetting
Catalog No. NC1772264 Supplier CELLTREAT Scientific Products Supplier No. 230300
CELLTREAT Scientific Products 2-20uL Adjustable Pipette is a high precision pipette with easy-to-read volume display. Continuously adjustable with locking rig to prevent volume changes while pipetting. Lightweight design and easy operation with either hand. Reduced operating forces: plunger is easy to press and dispense. Stainless steel and PTFE components for long life. It is not receommended to autocalve more than 6 times. Fits CELLTREAT Scientific Products Universal Pipette Tips for accurate pipetting